siRNA Details
| Gene Symbol: Bmpr1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024259.1 | NP_001019430.1 | 5 | 521 | ||
| XM_006233400.2 | XP_006233462.1 | 9 | 1365 | |||
| XM_006233401.2 | XP_006233463.1 | 8 | 1026 | |||
| XM_008761513.1 | XP_008759735.1 | 8 | 1225 | |||
| XM_008761514.1 | XP_008759736.1 | 11 | 1747 | |||
| GenBank mRNA | BC092609.1 | - | 5 | 521 | 7310757 | |

Bmpr1b
Genomic Map