siRNA Details
| Gene Symbol: Bmpr1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024259.1 | NP_001019430.1 | 6, 7 | 950 | ||
| XM_006233400.2 | XP_006233462.1 | 10, 11 | 1794 | |||
| XM_006233401.2 | XP_006233463.1 | 9, 10 | 1455 | |||
| XM_008761513.1 | XP_008759735.1 | 9, 10 | 1654 | |||
| XM_008761514.1 | XP_008759736.1 | 12, 13 | 2176 | |||
| GenBank mRNA | BC092609.1 | - | 6, 7 | 950 | 7310757 | |

Bmpr1b
Genomic Map