siRNA Details
| Gene Symbol: LOC310926 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025002.1 | NP_001020173.1 | 4 | 673 | ||
| GenBank mRNA | AY310143.1 | - | 4 | 673 | ||
| AY318956.1 | - | 4 | 673 | |||
| AY318962.1 | - | 4 | 673 | |||
| AY325183.1 | - | 4 | 673 | |||

LOC310926
Genomic Map