siRNA Details
Gene Symbol: Fpgt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199494.1 | NP_955788.1 | 4 | 1152 | ||
GenBank mRNA | AJ276066.1 | - | 4 | 1187 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Fpgt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199494.1 | NP_955788.1 | 4 | 1152 | ||
GenBank mRNA | AJ276066.1 | - | 4 | 1187 |