siRNA Details
| Gene Symbol: Plekhg5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_201272.1 | NP_958429.1 | 13 | 1645 | ||
| GenBank mRNA | AY499658.1 | - | 13 | 1645 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Plekhg5| Gene Symbol: Plekhg5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_201272.1 | NP_958429.1 | 13 | 1645 | ||
| GenBank mRNA | AY499658.1 | - | 13 | 1645 | ||