siRNA Details
| Gene Symbol: Nmi | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034148.1 | NP_001029320.1 | 260 | |||
| GenBank mRNA | BC100089.1 | - | 260 | 7371417 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nmi| Gene Symbol: Nmi | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034148.1 | NP_001029320.1 | 260 | |||
| GenBank mRNA | BC100089.1 | - | 260 | 7371417 | ||