siRNA Details
Gene Symbol: Kif5c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107730.1 | NP_001101200.1 | 2 | 559 | ||
XM_008761745.1 | XP_008759967.1 | 2 | 578 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Kif5c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107730.1 | NP_001101200.1 | 2 | 559 | ||
XM_008761745.1 | XP_008759967.1 | 2 | 578 |