siRNA Details
| Gene Symbol: Kif5c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107730.1 | NP_001101200.1 | 3 | 587 | ||
| XM_008761745.1 | XP_008759967.1 | 3 | 606 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kif5c| Gene Symbol: Kif5c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107730.1 | NP_001101200.1 | 3 | 587 | ||
| XM_008761745.1 | XP_008759967.1 | 3 | 606 | |||