siRNA Details
| Gene Symbol: Stam2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012085.1 | NP_001012085.1 | 3 | 246 | ||
| GenBank mRNA | BC083912.1 | - | 3 | 246 | 7134287 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Stam2| Gene Symbol: Stam2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012085.1 | NP_001012085.1 | 3 | 246 | ||
| GenBank mRNA | BC083912.1 | - | 3 | 246 | 7134287 | |