siRNA Details
| Gene Symbol: Galnt13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199106.1 | NP_954537.1 | 3 | 507 | ||
| XM_008761746.1 | XP_008759968.1 | 1 | 106 | |||
| GenBank mRNA | AY371923.1 | - | 3 | 507 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Galnt13| Gene Symbol: Galnt13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199106.1 | NP_954537.1 | 3 | 507 | ||
| XM_008761746.1 | XP_008759968.1 | 1 | 106 | |||
| GenBank mRNA | AY371923.1 | - | 3 | 507 | ||