siRNA Details
| Gene Symbol: Ccdc148 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107732.1 | NP_001101202.1 | 8 | 998 | ||
| XM_008761833.1 | XP_008760055.1 | 8 | 1084 | |||
| XM_008761834.1 | XP_008760056.1 | 8 | 1057 | |||
| XM_008761835.1 | XP_008760057.1 | 8 | 1581 | |||
| XM_008761836.1 | XP_008760058.1 | 8 | 1464 | |||
| XM_008761837.1 | XP_008760059.1 | 8 | 1084 | |||
| GenBank mRNA | BC167021.1 | - | 10 | 1269 | ||

Ccdc148
Genomic Map