siRNA Details
Gene Symbol: Tanc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002854.1 | NP_001002854.1 | 25 | 4185 | ||
XM_006234204.1 | XP_006234266.1 | 24 | 4104 | |||
XM_006234205.2 | XP_006234267.1 | 23 | 6340 | |||
XM_006234207.2 | XP_006234269.1 | 20 | 4302 | |||
XM_008761838.1 | XP_008760060.1 | 23 | 4518 | |||
XM_008761839.1 | XP_008760061.1 | 23 | 4804 | |||
XM_008761840.1 | XP_008760062.1 | 21 | 5436 | |||
GenBank mRNA | AB098072.1 | - | 25 | 4185 |
