siRNA Details
Gene Symbol: Tanc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002854.1 | NP_001002854.1 | 3 | 332 | ||
XM_006234204.1 | XP_006234266.1 | 2 | 230 | |||
GenBank mRNA | AB098072.1 | - | 3 | 332 |
