siRNA Details
| Gene Symbol: Zeb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033701.1 | NP_001028873.1 | 1683 | |||
| XM_006234144.2 | XP_006234206.2 | 2043 | ||||
| XM_006234145.2 | XP_006234207.1 | 2563 | ||||
| XM_006234147.2 | XP_006234209.1 | 7 | 2760 | |||
| XM_008761747.1 | XP_008759969.1 | 1809 | ||||
| XM_008761748.1 | XP_008759970.1 | 2042 | ||||
| GenBank mRNA | AM084708.1 | - | 1683 | |||

Zeb2
Genomic Map