siRNA Details
| Gene Symbol: Psmd14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025689.1 | NP_001020860.1 | 9 | 872 | ||
| GenBank mRNA | BC097427.1 | - | 9 | 872 | ||
| BC158645.1 | - | 9 | 820 | 8363877 | ||
| FQ228714.1 | - | 9 | 848 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Psmd14| Gene Symbol: Psmd14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025689.1 | NP_001020860.1 | 9 | 872 | ||
| GenBank mRNA | BC097427.1 | - | 9 | 872 | ||
| BC158645.1 | - | 9 | 820 | 8363877 | ||
| FQ228714.1 | - | 9 | 848 | |||