siRNA Details
| Gene Symbol: Xirp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_201989.1 | NP_973718.1 | 7 | 2147 | ||
| GenBank mRNA | AF466694.1 | - | 7 | 2147 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Xirp2| Gene Symbol: Xirp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_201989.1 | NP_973718.1 | 7 | 2147 | ||
| GenBank mRNA | AF466694.1 | - | 7 | 2147 | ||