siRNA Details
| Gene Symbol: Fastkd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191738.1 | NP_001178667.1 | 2, 3 | 1407 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fastkd1| Gene Symbol: Fastkd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191738.1 | NP_001178667.1 | 2, 3 | 1407 | ||