siRNA Details
| Gene Symbol: Fastkd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191738.1 | NP_001178667.1 | 4 | 1815 | ||
| XM_006234311.1 | XP_006234373.1 | 4 | 624 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fastkd1| Gene Symbol: Fastkd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191738.1 | NP_001178667.1 | 4 | 1815 | ||
| XM_006234311.1 | XP_006234373.1 | 4 | 624 | |||