siRNA Details
| Gene Symbol: Klhl23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134504.1 | NP_001127976.1 | 3 | 1430 | ||
| XM_006234313.2 | XP_006234375.1 | 3 | 1396 | |||
| XM_006234314.2 | XP_006234376.1 | 3 | 1892 | |||
| XM_006234316.2 | XP_006234378.1 | 3 | 1637 | |||

Klhl23
Genomic Map