siRNA Details
| Gene Symbol: RGD1565767 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_230013.6 | XP_230013.1 | 1 | 305 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1565767| Gene Symbol: RGD1565767 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_230013.6 | XP_230013.1 | 1 | 305 | ||