siRNA Details
| Gene Symbol: Osbpl6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107735.1 | NP_001101205.1 | 14 | 1778 | ||
| XM_008761900.1 | XP_008760122.1 | 12 | 1272 | |||
| XM_008761901.1 | XP_008760123.1 | 12 | 1272 | |||
| XM_008761902.1 | XP_008760124.1 | 11 | 1197 | |||
| XM_008761903.1 | XP_008760125.1 | 11 | 1179 | |||
| XM_008761904.1 | XP_008760126.1 | 11 | 1179 | |||
| XM_008761905.1 | XP_008760127.1 | 10 | 1104 | |||
| XM_008761906.1 | XP_008760128.1 | 10 | 1104 | |||
| XM_008761907.1 | XP_008760129.1 | 12 | 1272 | |||

Osbpl6
Genomic Map