siRNA Details
| Gene Symbol: Zfp385b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107736.1 | NP_001101206.1 | 4 | 1665 | ||
| XM_006234414.2 | XP_006234476.1 | 5 | 1324 | |||
| XM_006234416.2 | XP_006234478.1 | 4 | 1285 | |||
| XM_006234419.2 | XP_006234481.1 | 5 | 2481 | |||
| XM_006234420.2 | XP_006234482.1 | 5 | 850 | |||

Zfp385b
Genomic Map