siRNA Details
| Gene Symbol: Ssfa2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107738.1 | NP_001101208.1 | 11 | 2246 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ssfa2| Gene Symbol: Ssfa2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107738.1 | NP_001101208.1 | 11 | 2246 | ||