siRNA Details
| Gene Symbol: Rtn4rl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181380.2 | NP_852045.1 | 3 | 718 | ||
| XM_006234465.2 | XP_006234527.1 | 3 | 1030 | |||
| XM_006234466.2 | XP_006234528.1 | 3 | 1641 | |||
| XM_008761989.1 | XP_008760211.1 | 3 | 1213 | |||
| XM_008761990.1 | XP_008760212.1 | 3 | 818 | |||
| GenBank mRNA | AF532860.1 | - | 3 | 718 | ||
| AX751537.1 | - | 3 | 700 | |||

Rtn4rl2
Genomic Map