siRNA Details
| Gene Symbol: Slc43a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107743.1 | NP_001101213.1 | 6 | 796 | ||
| XM_006234467.2 | XP_006234529.2 | 6 | 810 | |||
| XM_006234468.2 | XP_006234530.2 | 6 | 722 | |||
| XM_006234470.2 | XP_006234532.1 | 6 | 669 | |||
| XM_008761991.1 | XP_008760213.1 | 6 | 674 | |||

Slc43a3
Genomic Map