siRNA Details
| Gene Symbol: Nup160 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107744.1 | NP_001101214.1 | 22, 23 | 3355 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nup160| Gene Symbol: Nup160 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107744.1 | NP_001101214.1 | 22, 23 | 3355 | ||