siRNA Details
| Gene Symbol: Fnbp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013159.2 | NP_001013177.2 | 8 | 1571 | ||
| XM_006234505.2 | XP_006234567.1 | 8 | 1571 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fnbp4| Gene Symbol: Fnbp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013159.2 | NP_001013177.2 | 8 | 1571 | ||
| XM_006234505.2 | XP_006234567.1 | 8 | 1571 | |||