siRNA Details
| Gene Symbol: Kbtbd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107746.1 | NP_001101216.1 | 3 | 725 | ||
| XM_006234507.2 | XP_006234569.1 | 3 | 717 | |||
| XM_006234508.1 | XP_006234570.1 | 3 | 807 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kbtbd4| Gene Symbol: Kbtbd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107746.1 | NP_001101216.1 | 3 | 725 | ||
| XM_006234507.2 | XP_006234569.1 | 3 | 717 | |||
| XM_006234508.1 | XP_006234570.1 | 3 | 807 | |||