siRNA Details
| Gene Symbol: Pex16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012088.1 | NP_001012088.1 | 7 | 733 | ||
| XM_006234567.2 | XP_006234629.1 | 7 | 839 | |||
| GenBank mRNA | BC089938.1 | - | 7 | 733 | 7310024 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pex16| Gene Symbol: Pex16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012088.1 | NP_001012088.1 | 7 | 733 | ||
| XM_006234567.2 | XP_006234629.1 | 7 | 839 | |||
| GenBank mRNA | BC089938.1 | - | 7 | 733 | 7310024 | |