siRNA Details
| Gene Symbol: Slc35c1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107748.1 | NP_001101218.1 | 2 | 1049 | ||
| XM_006234568.2 | XP_006234630.1 | 3 | 1258 | |||
| XM_008762026.1 | XP_008760248.1 | 3 | 1274 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc35c1| Gene Symbol: Slc35c1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107748.1 | NP_001101218.1 | 2 | 1049 | ||
| XM_006234568.2 | XP_006234630.1 | 3 | 1258 | |||
| XM_008762026.1 | XP_008760248.1 | 3 | 1274 | |||