siRNA Details
| Gene Symbol: Tp53i11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107749.1 | NP_001101219.1 | 6 | 505 | ||
| XM_006234569.2 | XP_006234631.1 | 6 | 733 | |||
| XM_006234570.2 | XP_006234632.1 | 6 | 694 | |||
| XM_006234571.2 | XP_006234633.1 | 5 | 360 | |||
| XM_006234572.2 | XP_006234634.1 | 6 | 543 | |||
| GenBank mRNA | BC167758.1 | - | 6 | 643 | 7123311 | |

Tp53i11
Genomic Map