siRNA Details
| Gene Symbol: Ext2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006234576.2 | XP_006234638.2 | 11 | 2026 | ||
| GenBank mRNA | BC166748.1 | - | 11 | 1984 | 5599208 | |
| FQ218956.1 | - | 2 | 276 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ext2| Gene Symbol: Ext2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006234576.2 | XP_006234638.2 | 11 | 2026 | ||
| GenBank mRNA | BC166748.1 | - | 11 | 1984 | 5599208 | |
| FQ218956.1 | - | 2 | 276 | |||