siRNA Details
| Gene Symbol: Ext2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107751.1 | NP_001101221.1 | 8 | 1440 | ||
| XM_006234576.2 | XP_006234638.2 | 8 | 1469 | |||
| GenBank mRNA | BC166748.1 | - | 8 | 1427 | 5599208 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ext2| Gene Symbol: Ext2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107751.1 | NP_001101221.1 | 8 | 1440 | ||
| XM_006234576.2 | XP_006234638.2 | 8 | 1469 | |||
| GenBank mRNA | BC166748.1 | - | 8 | 1427 | 5599208 | |