siRNA Details
| Gene Symbol: Nat10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107756.1 | NP_001101226.1 | 23 | 2584 | ||
| GenBank mRNA | BC169067.1 | - | 24 | 2692 | 7939299 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nat10| Gene Symbol: Nat10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107756.1 | NP_001101226.1 | 23 | 2584 | ||
| GenBank mRNA | BC169067.1 | - | 24 | 2692 | 7939299 | |