siRNA Details
| Gene Symbol: Knstrn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004264.1 | NP_001004264.1 | 6 | 710 | ||
| XM_006234757.2 | XP_006234819.1 | 6 | 746 | |||
| XM_006234758.2 | XP_006234820.1 | 6 | 567 | |||
| GenBank mRNA | BC079438.1 | - | 6 | 710 | 7115344 | |

Knstrn
Genomic Map