siRNA Details
| Gene Symbol: Rmdn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014046.2 | NP_001014068.1 | 2 | 251 | ||
| GenBank mRNA | BC082081.1 | - | 2 | 213 | 7133344 | |
| DN931790.1 | - | 2 | 283 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rmdn3| Gene Symbol: Rmdn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014046.2 | NP_001014068.1 | 2 | 251 | ||
| GenBank mRNA | BC082081.1 | - | 2 | 213 | 7133344 | |
| DN931790.1 | - | 2 | 283 | |||