siRNA Details
| Gene Symbol: Nusap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107762.1 | NP_001101232.1 | 5 | 772 | ||
| XM_006234768.2 | XP_006234830.1 | 5 | 603 | |||
| XM_006234769.2 | XP_006234831.1 | 5 | 603 | |||
| XM_006234770.2 | XP_006234832.1 | 4 | 456 | |||
| GenBank mRNA | BC161947.1 | - | 5 | 548 | 9025380 | |

Nusap1
Genomic Map