siRNA Details
| Gene Symbol: Ttbk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107766.1 | NP_001101236.1 | 14 | 3440 | ||
| XM_006234833.2 | XP_006234895.1 | 16 | 3873 | |||
| XM_006234834.2 | XP_006234896.1 | 16 | 3516 | |||
| XM_006234835.2 | XP_006234897.1 | 15 | 3799 | |||
| XM_006234836.2 | XP_006234898.1 | 14 | 3651 | |||
| XM_006234837.2 | XP_006234899.1 | 16 | 3519 | |||

Ttbk2
Genomic Map