siRNA Details
| Gene Symbol: Tmem62 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_230497.8 | XP_230497.4 | 13 | 2321 | ||
| GenBank mRNA | FQ210040.1 | - | 13 | 1725 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem62| Gene Symbol: Tmem62 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_230497.8 | XP_230497.4 | 13 | 2321 | ||
| GenBank mRNA | FQ210040.1 | - | 13 | 1725 | ||