siRNA Details
| Gene Symbol: Frmd5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008762211.1 | XP_008760433.1 | 1, 2 | 105 | ||
| XM_008762212.1 | XP_008760434.1 | 1, 2 | 105 | |||
| XM_008762213.1 | XP_008760435.1 | 1, 2 | 105 | |||
| XM_008762214.1 | XP_008760436.1 | 1, 2 | 133 | |||
| XM_008762215.1 | XP_008760437.1 | 1, 2 | 133 | |||

Frmd5
Genomic Map