siRNA Details
| Gene Symbol: Eif3j | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001077670.1 | NP_001071138.1 | 6 | 575 | ||
| GenBank mRNA | BC127509.1 | - | 6 | 575 | 7454886 | |
| FQ226401.1 | - | 6 | 524 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Eif3j| Gene Symbol: Eif3j | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001077670.1 | NP_001071138.1 | 6 | 575 | ||
| GenBank mRNA | BC127509.1 | - | 6 | 575 | 7454886 | |
| FQ226401.1 | - | 6 | 524 | |||