siRNA Details
| Gene Symbol: Spg11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191571.1 | NP_001178500.1 | 5 | 913 | ||
| XM_006234855.2 | XP_006234917.1 | 5 | 1268 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Spg11| Gene Symbol: Spg11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191571.1 | NP_001178500.1 | 5 | 913 | ||
| XM_006234855.2 | XP_006234917.1 | 5 | 1268 | |||