siRNA Details
| Gene Symbol: Sppl2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107770.1 | NP_001101240.1 | 15 | 1732 | ||
| XM_006234920.2 | XP_006234982.1 | 16 | 1647 | |||
| GenBank mRNA | FQ210986.1 | - | 13 | 1178 | ||
| FQ226494.1 | - | 7 | 513 | |||

Sppl2a
Genomic Map