siRNA Details
| Gene Symbol: Zc3h8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012090.1 | NP_001012090.1 | 4 | 522 | ||
| GenBank mRNA | BC079122.1 | - | 4 | 522 | 7126707 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zc3h8| Gene Symbol: Zc3h8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012090.1 | NP_001012090.1 | 4 | 522 | ||
| GenBank mRNA | BC079122.1 | - | 4 | 522 | 7126707 | |