siRNA Details
| Gene Symbol: Stk35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107773.1 | NP_001101243.1 | 3 | 2259 | ||
| XM_006234974.2 | XP_006235036.1 | 3 | 1207 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Stk35| Gene Symbol: Stk35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107773.1 | NP_001101243.1 | 3 | 2259 | ||
| XM_006234974.2 | XP_006235036.1 | 3 | 1207 | |||