siRNA Details
| Gene Symbol: Kiz | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006235181.2 | XP_006235243.1 | 7 | 1498 | ||
| XM_006235182.2 | XP_006235244.1 | 7 | 1498 | |||
| XM_008762291.1 | XP_008760513.1 | 7 | 1498 | |||
| XM_008762292.1 | XP_008760514.1 | 4 | 1269 | |||

Kiz
Genomic Map