siRNA Details
| Gene Symbol: Gzf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107788.1 | NP_001101258.1 | 2 | 373 | ||
| XM_006235153.2 | XP_006235215.1 | 2 | 240 | |||
| XM_006235154.2 | XP_006235216.1 | 2 | 1352 | |||
| XM_008762274.1 | XP_008760496.1 | 3 | 531 | |||

Gzf1
Genomic Map