siRNA Details
| Gene Symbol: Nanp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009409.1 | NP_001009409.1 | 1, 2 | 104 | ||
| GenBank mRNA | BC087587.1 | - | 1, 2 | 105 | 7319178 | |
| CO399358.1 | - | 1, 2 | 104 | |||
| CO568277.1 | - | 1, 2 | 83 | |||

Nanp
Genomic Map