siRNA Details
| Gene Symbol: Fam110a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014050.2 | NP_001014072.2 | 2 | 1344 | ||
| NM_001244768.1 | NP_001231697.1 | 2 | 1225 | |||
| NM_001244769.1 | NP_001231698.1 | 2 | 1297 | |||
| NM_001244770.1 | NP_001231699.1 | 2 | 1144 | |||
| XM_006235231.2 | XP_006235293.1 | 2 | 1597 | |||
| XM_006235234.2 | XP_006235296.1 | 3 | 1332 | |||
| GenBank mRNA | BC079251.1 | - | 2 | 1344 | 7133086 | |

Fam110a
Genomic Map