siRNA Details
| Gene Symbol: Itch | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005887.1 | NP_001005887.1 | 17 | 1831 | ||
| XM_008762335.1 | XP_008760557.1 | 19 | 2059 | |||
| XM_008762336.1 | XP_008760558.1 | 19 | 1984 | |||
| XM_008762337.1 | XP_008760559.1 | 16 | 1736 | |||
| GenBank mRNA | AY600518.1 | - | 17 | 1831 | ||

Itch
Genomic Map